CAS RN 959-88-6
IUPAC Name:
1-bromo-4-[4-(4-bromophenyl)buta-1,3-diyn-1-yl]benzene
Synonyms:
1-bromo-4-[4-(4-bromophenyl)buta-1,3-diyn-1-yl]benzene
Molecular Weight (g/mol):
360.05
Molecular Formula:
C16H8Br2
InChi Key:
IIERBCCHBFZMQZ-UHFFFAOYSA-N
SMILES:
BrC1=CC=C(C=C1)C#CC#CC1=CC=C(Br)C=C1
1
–
1
of
1
results
1
1,4-Bis(4-bromophenyl)-1,3-butadiyne 98.0+%, TCI America™
CAS: 959-88-6 Molecular Formula: C16H8Br2 Molecular Weight (g/mol): 360.05 MDL Number: MFCD00825678 InChI Key: IIERBCCHBFZMQZ-UHFFFAOYSA-N Synonym: 1,4-bis 4-bromophenyl-1,3-butadiyne,1,3-butadiyne, 1,4-di 4-bromophenyl,4,4'-dibromodiphenylbutadiyne,1,4-bis 4-bromophenyl buta-1,3-diyne,1,1'-buta-1,3-diyne-1,4-diyl bis 4-bromobenzene,1-bromo-4-4-4-bromophenyl-1,3-butadiynyl benzene,1-bromo-4-4-4-bromophenyl buta-1,3-diynyl benzene,1-bromo-4-4-4-bromophenyl-1,3-butadiynyl benzene #,1-bromo-4-4-4-bromophenyl buta-1,3-diyn-1-yl benzene PubChem CID: 632304 IUPAC Name: 1-bromo-4-[4-(4-bromophenyl)buta-1,3-diyn-1-yl]benzene SMILES: BrC1=CC=C(C=C1)C#CC#CC1=CC=C(Br)C=C1