Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,4-Bis(4-bromophenyl)-1,3-butadiyne 98.0+%, TCI America™
SDP

Catalog No. B4013200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B4013200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B4013200MG Supplier TCI America Supplier No. B4013200MG

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 959-88-6
Molecular Formula C16H8Br2
Molecular Weight (g/mol) 360.05
MDL Number MFCD00825678
InChI Key IIERBCCHBFZMQZ-UHFFFAOYSA-N
Synonym 1,4-bis 4-bromophenyl-1,3-butadiyne, 1,3-butadiyne, 1,4-di 4-bromophenyl, 4,4'-dibromodiphenylbutadiyne, 1,4-bis 4-bromophenyl buta-1,3-diyne, 1,1'-buta-1,3-diyne-1,4-diyl bis 4-bromobenzene, 1-bromo-4-4-4-bromophenyl-1,3-butadiynyl benzene, 1-bromo-4-4-4-bromophenyl buta-1,3-diynyl benzene, 1-bromo-4-4-4-bromophenyl-1,3-butadiynyl benzene #, 1-bromo-4-4-4-bromophenyl buta-1,3-diyn-1-yl benzene
PubChem CID 632304
IUPAC Name 1-bromo-4-[4-(4-bromophenyl)buta-1,3-diyn-1-yl]benzene
SMILES BrC1=CC=C(C=C1)C#CC#CC1=CC=C(Br)C=C1

Specifications

Melting Point 265°C
Color Yellow
Quantity 200 mg
Formula Weight 360.05
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 1,4-Bis(4-bromophenyl)-1,3-butadiyne
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.