accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClClCl(R)OSOO(S)(R)ClClCl

CAS RN 959-98-8

IUPAC Name: (1R,2S,8R)-1,9,10,11,12,12-hexachloro-4,6-dioxa-5λ⁴-thiatricyclo[7.2.1.0²,⁸]dodec-10-en-5-one
Synonyms: (1R,2S,8R)-1,9,10,11,12,12-hexachloro-4,6-dioxa-5λ⁴-thiatricyclo[7.2.1.0²,⁸]dodec-10-en-5-one
Molecular Weight (g/mol): 406.9
Molecular Formula: C9H6Cl6O3S
InChi Key: RDYMFSUJUZBWLH-GDSHQCHSSA-N
SMILES: ClC1=C(Cl)[C@]2(Cl)[C@@H]3COS(=O)OC[C@@H]3C1(Cl)C2(Cl)Cl
Element 
  • (1)
  • (1)
  • (1)
Grade 
  • (1)
Solvent or Matrix 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)