CAS RN 95950-70-2
IUPAC Name:
2,6-diphenylanthracene
Synonyms:
2,6-diphenylanthracene
Molecular Weight (g/mol):
330.43
Molecular Formula:
C26H18
InChi Key:
MZBIWFMZBZJUHX-UHFFFAOYSA-N
SMILES:
C1=CC=C(C=C1)C1=CC2=CC3=CC=C(C=C3C=C2C=C1)C1=CC=CC=C1