Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Diphenylanthracene (purified by sublimation) 98.0+%, TCI America™
SDP

Catalog No. D5152100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D5152100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D5152100MG Supplier TCI America Supplier No. D5152100MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 95950-70-2
Molecular Formula C26H18
Molecular Weight (g/mol) 330.43
MDL Number MFCD30721942
InChI Key MZBIWFMZBZJUHX-UHFFFAOYSA-N
PubChem CID 14915456
IUPAC Name 2,6-diphenylanthracene
SMILES C1=CC=C(C=C1)C1=CC2=CC3=CC=C(C=C3C=C2C=C1)C1=CC=CC=C1

Specifications

Color Yellow
Quantity 100 mg
Formula Weight 330.43
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,6-Diphenylanthracene (purified by sublimation)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.