CAS RN 959626-45-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ cFMS Receptor Inhibitor IV
The cFMS Receptor Inhibitor IV, also referenced under CAS 959626-45-0, controls the biological activity of cFMS. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.