Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ cFMS Receptor Inhibitor IV

Catalog No. 3440395MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
34-403-95MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 34-403-95MG Supplier MilliporeSigma™ Supplier No. 3440395MG
Add to Cart
Add to Cart

The cFMS Receptor Inhibitor IV, also referenced under CAS 959626-45-0, controls the biological activity of cFMS. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Packaged Under Inert Gas
TRUSTED_SUSTAINABILITY

Specifications

Color Yellow
Description 95% by HPLC
CAS 959626-45-0
Molecular Formula C22H26N4O2
Formula Weight 378.5g/mol
Solubility DMSO
Content And Storage Protect from light
Form Solid
Quantity 5 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.