CAS RN 959979-30-7
CAS RN 959979-30-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Promotions Available
Sigma Aldrich 1,2-Bis[(2R,5R)-2,5-Dimethylphospholano]-3,3,4,4-Tetrafluoro-1-Cyclobutene(1,5-Cyclooctadiene)Rhodium(I) Tetrafluoroborate
Sigma Aldrich 1,2-Bis[(2R,5R)-2,5-Dimethylphospholano]-3,3,4,4-Tetrafluoro-1-Cyclobutene(1,5-Cyclooctadiene)Rhodium(I) Tetrafluoroborate
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.