accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 96-09-3

IUPAC Name: 2-phenyloxirane
Synonyms: (+-)-styrene oxide 1-phenyloxirane styrene oxide, (s)-isomer styrene oxide, (+-)-isomer styrene epoxide (s)-isomer styrene oxide styrene-7,8-oxide (+-)-isomer styrene oxide styrene oxide, (R)-isomer (epoxyethyl)benzene (styrene oxide)
Molecular Weight (g/mol): 120.15
Molecular Formula: C8H8O
InChi Key: AWMVMTVKBNGEAK-UHFFFAOYNA-N
SMILES: C1OC1C1=CC=CC=C1
Boiling Point 
  • (2)
  • (5)
  • (4)
Molecular Weight (g/mol) 
  • (7)
  • (4)
Percent Purity 
  • (2)
  • (2)
  • (3)
  • (4)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (3)
  • (2)
  • (6)
  • (2)
  • (5)
  • (4)
  • (7)
  • (4)
  • (2)
  • (2)
  • (3)
  • (4)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (6)