accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClClCl

CAS RN 96-18-4

IUPAC Name: 1,2,3-trichloropropane
Synonyms: 1,2,3-trichloropropane C3H5cl3 trichlorohydrin glyceryl trichlorohydrin glycerol trichlorohydrin
Molecular Weight (g/mol): 147.42
Molecular Formula: C3H5Cl3
InChi Key: CFXQEHVMCRXUSD-UHFFFAOYSA-N
SMILES: ClCC(Cl)CCl
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
  • (2)
  • (2)
Boiling Point 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (1)