Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2,3-Trichloropropane, MP Biomedicals

Catalog No. ICN219119 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN219119 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN219119 Supplier MP Biomedicals, Inc Supplier No. 0521911980

Chemical Identifiers

CAS 96-18-4
Molecular Formula C3H5Cl3
Molecular Weight (g/mol) 147.42
MDL Number MFCD00000946
InChI Key CFXQEHVMCRXUSD-UHFFFAOYSA-N
Synonym trichlorohydrin, allyl trichloride, propane, 1,2,3-trichloro, glycerol trichlorohydrin, glyceryl trichlorohydrin, unii-3mj7qck0z0, ccris 5874, 3mj7qck0z0, dsstox_cid_1390, dsstox_rid_76132
PubChem CID 7285
ChEBI CHEBI:34036
IUPAC Name 1,2,3-trichloropropane
SMILES ClCC(Cl)CCl

Specifications

Quantity 100 g
Formula Weight 147.44
Chemical Name or Material 1, 2, 3-Trichloropropane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.