accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClClCl(Z)

CAS RN 96-19-5

IUPAC Name: (1Z)-1,2,3-trichloroprop-1-ene
Synonyms: 1,2,3-trichloropropene
Molecular Weight (g/mol): 145.41
Molecular Formula: C3H3Cl3
InChi Key: HIILBTHBHCLUER-IWQZZHSRSA-N
SMILES: ClC\C(Cl)=C\Cl
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results