Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2,3-Trichloropropene (cis- and trans- mixture) 95.0+%, TCI America™
SDP

Catalog No. T0396500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T0396500G 500 g
T039625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T0396500G Supplier TCI America Supplier No. T0396500G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 96-19-5
Molecular Formula C3H3Cl3
Molecular Weight (g/mol) 145.41
MDL Number MFCD00018931
InChI Key HIILBTHBHCLUER-IWQZZHSRSA-N
PubChem CID 5370144
IUPAC Name (1Z)-1,2,3-trichloroprop-1-ene
SMILES ClC\C(Cl)=C\Cl

Specifications

Color Yellow
Boiling Point 145°C
Quantity 500 g
UN Number 2810
Formula Weight 145.41
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,2,3-Trichloropropene (cis- and trans- mixture)
RTECSNumber UD2450000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.