accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NCH3ONOONOOSH

CAS RN 96034-64-9

IUPAC Name: (4-nitrophenyl)methyl 2-(dimethylcarbamoyl)-4-sulfanylpyrrolidine-1-carboxylate
Synonyms: (4-nitrophenyl)methyl 2-(dimethylcarbamoyl)-4-sulfanylpyrrolidine-1-carboxylate
Molecular Weight (g/mol): 353.39
Molecular Formula: C15H19N3O5S
InChi Key: VGLBNJWGUYQZHD-UHFFFAOYNA-N
SMILES: CN(C)C(=O)C1CC(S)CN1C(=O)OCC1=CC=C(C=C1)[N+]([O-])=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results