Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Nitrobenzyl (2S,4S)-2-(Dimethylcarbamoyl)-4-mercapto-1-pyrrolidinecarboxylate 95.0+%, TCI America™
SDP

Catalog No. N08281G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N08281G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N08281G Supplier TCI America Supplier No. N08281G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 96034-64-9
Molecular Formula C15H19N3O5S
Molecular Weight (g/mol) 353.393
MDL Number MFCD08063348
InChI Key VGLBNJWGUYQZHD-STQMWFEESA-N
Synonym (2S,4S)-2-(Dimethylcarbamoyl)-4-mercapto-1-pyrrolidinecarboxylic Acid 4-Nitrobenzyl Ester
PubChem CID 10871907
IUPAC Name (4-nitrophenyl)methyl (2S,4S)-2-(dimethylcarbamoyl)-4-sulfanylpyrrolidine-1-carboxylate
SMILES CN(C)C(=O)C1CC(CN1C(=O)OCC2=CC=C(C=C2)[N+](=O)[O-])S

Specifications

Melting Point 119°C
Color White-Yellow
Quantity 1 g
Formula Weight 353.39
Percent Purity ≥95.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Nitrobenzyl (2S,4S)-2-(Dimethylcarbamoyl)-4-mercapto-1-pyrrolidinecarboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.