accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 961-07-9

IUPAC Name: 2-amino-9-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-6,9-dihydro-1H-purin-6-one
Synonyms: 2-deoxyguanosine
Molecular Weight (g/mol): 267.25
Molecular Formula: C10H13N5O4
InChi Key: YKBGVTZYEHREMT-DWFKHXNSNA-N
SMILES: NC1=NC2=C(N=CN2[C@H]2C[C@H](O)[C@@H](CO)O2)C(=O)N1
Physical Form 
  • (5)
  • (3)
  • (1)
Quantity 
  • (3)
  • (1)
  • (1)
  • (1)
  • (3)
Percent Purity 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (5)
  • (5)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)
  • (3)