accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 961-27-3

IUPAC Name: 1-(7-acetyl-9H-fluoren-2-yl)ethan-1-one
Synonyms: 1-(7-acetyl-9H-fluoren-2-yl)ethanone
Molecular Weight (g/mol): 250.3
Molecular Formula: C17H14O2
InChi Key: RIRYGERFWHUZBT-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC=C2C(CC3=CC(=CC=C23)C(C)=O)=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results