Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,7-Diacetylfluorene 98.0+%, TCI America™
SDP

Catalog No. D28631G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D28631G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D28631G Supplier TCI America Supplier No. D28631G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 961-27-3
Molecular Formula C17H14O2
Molecular Weight (g/mol) 250.30
MDL Number MFCD00045332
InChI Key RIRYGERFWHUZBT-UHFFFAOYSA-N
Synonym 2,7-diacetylfluorene, 1-7-acetyl-9h-fluoren-2-yl ethan-1-one, 1-7-acetyl-9h-fluoren-2-yl ethanone, 1,1'-9h-fluorene-2,7-diyl diethanone, 2,7-diacetyl fluorene, 9h-fluoren-9-one,2,7-diacetyl, ethanone,1,1'-9h-fluorene-2,7-diyl bis, 2,7-diacetyl-fluorene, 2.7-diacetylfluoren, 2 7-diacetyl fluorene
PubChem CID 283001
IUPAC Name 1-(7-acetyl-9H-fluoren-2-yl)ethan-1-one
SMILES CC(=O)C1=CC=C2C(CC3=CC(=CC=C23)C(C)=O)=C1

Specifications

Color Yellow
Quantity 1 g
Formula Weight 250.30
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,7-Diacetylfluorene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.