CAS RN 96507-86-7
IUPAC Name:
1,2-bis(9-methyldecyl) benzene-1,2-dicarboxylate
Synonyms:
1,2-bis(9-methyldecyl) phthalate
Molecular Weight (g/mol):
474.73
Molecular Formula:
C30H50O4
InChi Key:
LGBAGUMSAPUZPU-UHFFFAOYSA-N
SMILES:
CC(C)CCCCCCCCOC(=O)C1=CC=CC=C1C(=O)OCCCCCCCCC(C)C
1
–
1
of
1
results
1
Diundecyl Phthalate (mixture of branched chain isomers) 98.0+%, TCI America™
CAS: 96507-86-7 Molecular Formula: C30H50O4 Molecular Weight (g/mol): 474.726 InChI Key: LGBAGUMSAPUZPU-UHFFFAOYSA-N Synonym: Phthalic Acid Diundecyl Ester PubChem CID: 175686 IUPAC Name: bis(9-methyldecyl) benzene-1,2-dicarboxylate SMILES: CC(C)CCCCCCCCOC(=O)C1=CC=CC=C1C(=O)OCCCCCCCCC(C)C