CAS RN 96744-75-1
IUPAC Name:
N-methyl-2-phenyl-N-[(5R,7S,8S)-7-(pyrrolidin-1-yl)-1-oxaspiro[4.5]decan-8-yl]acetamide
Synonyms:
N-methyl-2-phenyl-N-[(5R,7S,8S)-7-(pyrrolidin-1-yl)-1-oxaspiro[4.5]decan-8-yl]acetamide
Molecular Weight (g/mol):
356.51
Molecular Formula:
C22H32N2O2
InChi Key:
PGZRDDYTKFZSFR-ONTIZHBOSA-N
SMILES:
CN([C@H]1CC[C@@]2(CCCO2)C[C@@H]1N1CCCC1)C(=O)CC1=CC=CC=C1
1
–
1
of
1
results
1
Promotions Available
Thermo Scientific Chemicals U-69593, 99%
Thermo Scientific Chemicals U-69593, 99%
CAS: 96744-75-1 Molecular Formula: C22H32N2O2