accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 96761-85-2

IUPAC Name: 3'-bromo-3,5-bis(3-bromophenyl)-1,1'-biphenyl
Synonyms: 3'-bromo-3,5-bis(3-bromophenyl)-1,1'-biphenyl
Molecular Weight (g/mol): 543.1
Molecular Formula: C24H15Br3
InChi Key: JKCQADHKVQXKFF-UHFFFAOYSA-N
SMILES: BrC1=CC=CC(=C1)C1=CC(=CC(=C1)C1=CC(Br)=CC=C1)C1=CC(Br)=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results