Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3,5-Tris(3-bromophenyl)benzene 96.0+%, TCI America™
SDP

Catalog No. T30735G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T30735G 5 g
T30731G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T30735G Supplier TCI America Supplier No. T30735G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 96761-85-2
Molecular Formula C24H15Br3
Molecular Weight (g/mol) 543.096
MDL Number MFCD02323390
InChI Key JKCQADHKVQXKFF-UHFFFAOYSA-N
PubChem CID 4576280
IUPAC Name 1,3,5-tris(3-bromophenyl)benzene
SMILES C1=CC(=CC(=C1)Br)C2=CC(=CC(=C2)C3=CC(=CC=C3)Br)C4=CC(=CC=C4)Br

Specifications

Melting Point 172°C
Color White-Yellow
Quantity 5 g
Formula Weight 543.10
Percent Purity ≥96.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 1,3,5-Tris(3-bromophenyl)benzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.