accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 97-02-9

IUPAC Name: 2,4-dinitroaniline
Synonyms: 2,4-dinitroaniline 2,4-dinitroanilina 2,4-nitroaniline 2,4-dinitroaminobenzene
Molecular Weight (g/mol): 183.12
Molecular Formula: C6H5N3O4
InChi Key: LXQOQPGNCGEELI-UHFFFAOYSA-N
SMILES: NC1=CC=C(C=C1[N+]([O-])=O)[N+]([O-])=O
Quantity 
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (4)
Percent Purity 
  • (2)
  • (2)
  • (1)
  • (6)
Physical Form 
  • (7)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (1)
  • (1)
  • (4)
  • (6)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (4)
  • (2)
  • (2)
  • (1)
  • (6)
  • (7)