Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4-Dinitroaniline, MP Biomedicals

Catalog No. ICN204322 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN204322 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN204322 Supplier MP Biomedicals, Inc Supplier No. 0520432205

Chemical Identifiers

CAS 97-02-9
Molecular Formula C6H5N3O4
Molecular Weight (g/mol) 183.12
MDL Number MFCD00007151
InChI Key LXQOQPGNCGEELI-UHFFFAOYSA-N
Synonym benzenamine, 2,4-dinitro, 1-amino-2,4-dinitrobenzene, 2,4-dinitroanilin, 2,4-dinitrobenzenamine, 2,4-dinitroanilina, 2,4-dinitraniline, 2,4-dinitrophenylamine, aniline, 2,4-dinitro, 2,4-dinitroaminobenzene, 2,4-nitroaniline
PubChem CID 7321
ChEBI CHEBI:34242
IUPAC Name 2,4-dinitroaniline
SMILES NC1=CC=C(C=C1[N+]([O-])=O)[N+]([O-])=O

Specifications

Quantity 5 g
Formula Weight 183.12
Chemical Name or Material 2, 4-Dinitroaniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.