accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHClClSClClHO

CAS RN 97-18-7

IUPAC Name: 2,4-dichloro-6-[(3,5-dichloro-2-hydroxyphenyl)sulfanyl]phenol
Synonyms: bithionol bisoxyphen actamer caswell no. 852 bithionol sulfide bithionol (substance) bithin bithional tkhsd TBP
Molecular Weight (g/mol): 356.04
Molecular Formula: C12H6Cl4O2S
InChi Key: JFIOVJDNOJYLKP-UHFFFAOYSA-N
SMILES: OC1=C(Cl)C=C(Cl)C=C1SC1=CC(Cl)=CC(Cl)=C1O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results