Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2'-Thiobis(4,6-dichlorophenol) 97.0+%, TCI America™
SDP

Catalog No. T0865500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T0865500G 500 g
T086525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T0865500G Supplier TCI America Supplier No. T0865500G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 97-18-7
Molecular Formula C12H6Cl4O2S
Molecular Weight (g/mol) 356.038
MDL Number MFCD00055727
InChI Key JFIOVJDNOJYLKP-UHFFFAOYSA-N
Synonym Bis(3,5-dichloro-2-hydroxyphenyl) Sulfide, Bithionol
PubChem CID 2406
ChEBI CHEBI:3131
IUPAC Name 2,4-dichloro-6-(3,5-dichloro-2-hydroxyphenyl)sulfanylphenol
SMILES C1=C(C=C(C(=C1Cl)O)SC2=CC(=CC(=C2O)Cl)Cl)Cl

Specifications

Melting Point 187°C
Color White-Yellow
Quantity 500 g
UN Number 2811
Formula Weight 356.04
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,2′-Thiobis(4,6-dichlorophenol)
RTECSNumber SN0525000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.