accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 97-78-9

IUPAC Name: 2-(N-methyldodecanamido)acetic acid
Synonyms: gardol
Molecular Weight (g/mol): 271.4
Molecular Formula: C15H29NO3
InChi Key: BACYUWVYYTXETD-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCC(=O)N(C)CC(O)=O
Molecular Weight (g/mol) 
  • (2)
  • (6)
Quantity 
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
Percent Purity 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results