Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Lauroylsarcosine 90.0+%, TCI America™
SDP

Catalog No. L0151500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L0151500G 500 g
L015125G 25 g
L0151100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. L0151500G Supplier TCI America Supplier No. L0151500G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 97-78-9
Molecular Formula C15H29NO3
Molecular Weight (g/mol) 271.40
MDL Number MFCD00021749
InChI Key BACYUWVYYTXETD-UHFFFAOYSA-N
Synonym n-lauroylsarcosine, lauroylsarcosine, n-dodecanoylsarcosine, lauroyl sarcosine, hamposyl l, maprosyl l, sarcosyl l, n-laurylsarcosine, n-dodecanoyl-n-methylglycine, glycine, n-methyl-n-1-oxododecyl
PubChem CID 7348
IUPAC Name 2-(N-methyldodecanamido)acetic acid
SMILES CCCCCCCCCCCC(=O)N(C)CC(O)=O

Specifications

Melting Point 46°C
Quantity 500 g
Formula Weight 271.40
Percent Purity ≥90.0% (GC)
Chemical Name or Material N-Lauroylsarcosine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.