accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOO

CAS RN 97-99-4

IUPAC Name: (oxolan-2-yl)methanol
Synonyms: tetrahydrofurfuryl alcohol
Molecular Weight (g/mol): 102.13
Molecular Formula: C5H10O2
InChi Key: BSYVTEYKTMYBMK-UHFFFAOYNA-N
SMILES: OCC1CCCO1
Percent Purity 
  • (2)
  • (6)
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (8)
  • (3)
  • (2)
  • (8)
  • (2)
  • (6)
  • (2)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)