Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Tetrahydrofurfuryl alcohol

Catalog No. T126533KG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
50 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T126533KG 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T126533KG Supplier Sigma Aldrich Supplier No. T126533KG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

The product is an 80wt.% solution of propargyl bromide (3-bromo-1-propyne) in xylene.Propargyl bromide solution (80wt.% solution in xylene) may be used in the preparation of dimethyl 5-hydroxyisophthalate propargyl ether.

Specifications

CAS 97-99-4
Melting Point -80°C (lit.)
Density 1.054 g/mL (at 25°C (literature))
Boiling Point 178°C (lit.)
Molecular Formula C5H10O2
Refractive Index n20/D 1.452 (literature)
Linear Formula C5H10O2
MDL Number MFCD00005372
Quantity 50 g
Synonym Oxolan-2-yl methanol; THFA; NSC 15434; Tetrahydro-2-furanmethanol
Molecular Weight (g/mol) 102.13
Percent Purity 98%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.