accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 97033-27-7

NClNCl

CAS RN 97033-27-7

IUPAC Name: 2,2'-dichloro-3,3'-bipyridine
Synonyms: 2,2'-dichloro-3,3'-bipyridine
Molecular Weight (g/mol): 225.07
Molecular Formula: C10H6Cl2N2
InChi Key: UGCBXWMUKMWBSV-UHFFFAOYSA-N
SMILES: ClC1=C(C=CC=N1)C1=C(Cl)N=CC=C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results