accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 97240-79-4

IUPAC Name: [(1R,2S,6S,9R)-4,4,11,11-tetramethyl-3,5,7,10,12-pentaoxatricyclo[7.3.0.0²,⁶]dodecan-6-yl]methyl sulfamate
Synonyms: topiramate topamax topamax sprinkle ortho brand of topiramate epitomax topiramic acid TPM topiramate (product) tipiramato tipiramate
Molecular Weight (g/mol): 339.36
Molecular Formula: C12H21NO8S
InChi Key: KJADKKWYZYXHBB-XBWDGYHZSA-N
SMILES: CC1(C)O[C@@H]2CO[C@@]3(COS(N)(=O)=O)OC(C)(C)O[C@H]3[C@@H]2O1
Molecular Weight (g/mol) 
  • (2)
  • (6)
Percent Purity 
  • (2)
  • (4)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
Grade 
  • (1)
  • (4)
Physical Form 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (4)
  • (1)
  • (4)
  • (2)
  • (2)
  • (4)
  • (2)
  • (6)
  • (2)
  • (4)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (4)
  • (2)
  • (3)