Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Topiramate, USP, 98-102%, Spectrum™ Chemical
SDP

Catalog No. 18607471 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
1 kg
Packaging:
Amber Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-607-471 1 kg Amber Glass Bottle
18-607-468 5 g Amber Glass Bottle
18-607-469 25 g Amber Glass Bottle
18-607-470 100 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-607-471 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. T16961KGBL
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

T1696, 97240-79-4, C12H21NO8S

Spectrum™ Chemical Topiramate, USP is used alone or with other medications to prevent and control seizures like epilepsy. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 97240-79-4
Molecular Formula C12H21NO8S
Molecular Weight (g/mol) 339.36
InChI Key KJADKKWYZYXHBB-XBWDGYHZSA-N
IUPAC Name [(1R,2S,6S,9R)-4,4,11,11-tetramethyl-3,5,7,10,12-pentaoxatricyclo[7.3.0.02,6]dodecan-6-yl]methyl sulfamate
SMILES CC1(C)O[C@@H]2CO[C@@]3(COS(N)(=O)=O)OC(C)(C)O[C@H]3[C@@H]2O1

Specifications

CAS Max % 1
Ignition Residue 0.002
Packaging Amber Glass Bottle
Quantity 1 kg
Specific Rotation -28.6° to -35.0°
Percent Purity 98 to 102%
Grade USP
Chemical Name or Material Topiramate
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.