CAS RN 97345-90-9
CAS RN 97345-90-9
IUPAC Name:
1,3-dimethyl-1H-imidazol-3-ium methyl sulfate
Synonyms:
1,3-dimethylimidazolium methyl sulfate(1-)
Molecular Weight (g/mol):
208.23
Molecular Formula:
C6H12N2O4S
InChi Key:
WOKQGMYCUGJNIJ-UHFFFAOYSA-M
SMILES:
COS([O-])(=O)=O.CN1C=C[N+](C)=C1
1
–
1
of
1
results
1
1,3-Dimethylimidazolium Methyl Sulfate 98.0+%, TCI America™
CAS: 97345-90-9 Molecular Formula: C6H12N2O4S Molecular Weight (g/mol): 208.232 MDL Number: MFCD03453276 InChI Key: WOKQGMYCUGJNIJ-UHFFFAOYSA-M PubChem CID: 12075598 IUPAC Name: 1,3-dimethylimidazol-1-ium;methyl sulfate SMILES: CN1C=C[N+](=C1)C.COS(=O)(=O)[O-]