Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Dimethylimidazolium Methyl Sulfate 98.0+%, TCI America™
SDP

Catalog No. D53485G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D53485G 5 g
D534825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D53485G Supplier TCI America Supplier No. D53485G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 97345-90-9
Molecular Formula C6H12N2O4S
Molecular Weight (g/mol) 208.232
MDL Number MFCD03453276
InChI Key WOKQGMYCUGJNIJ-UHFFFAOYSA-M
PubChem CID 12075598
IUPAC Name 1,3-dimethylimidazol-1-ium;methyl sulfate
SMILES CN1C=C[N+](=C1)C.COS(=O)(=O)[O-]

Specifications

Quantity 5 g
Synonym 1,3-dimethylimidazolium methylsulfate, 1,3-dimethylimidazolium methyl sulfate, 1,3-dimethyl-1h-imidazol-3-ium methyl sulfate, 1,3-dimethylimidazolium methyl sulfate 1-, dmimmeso4, 1, 3-dimethylimidazolium methylsulfate, 1,3-dimethylimidazolium methyl sulfate, for catalysis hplc
Formula Weight 208.23
Percent Purity ≥98.0% (HPLC,N)
Physical Form Liquid
Chemical Name or Material 1,3-Dimethylimidazolium Methyl Sulfate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.