accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CHNOHH2NOOHOOH(E/Z)

CAS RN 97469-12-0

IUPAC Name: 3-(2-aminopropyl)-1H-indol-5-ol; but-2-enedioic acid
Synonyms: α-methylserotonin
Molecular Weight (g/mol): 306.32
Molecular Formula: C15H18N2O5
InChi Key: YQNHFSXRABPJLP-UHFFFAOYNA-N
SMILES: OC(=O)C=CC(O)=O.CC(N)CC1=CNC2=CC=C(O)C=C12
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results