accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 97538-67-5

OONClClCl

CAS RN 97538-67-5

IUPAC Name: 3-(trichloromethyl)-hexahydropyrrolo[1,2-c][1,3]oxazol-1-one
Synonyms: 3-(trichloromethyl)-tetrahydro-3H-pyrrolo[1,2-c][1,3]oxazol-1-one
Molecular Weight (g/mol): 244.5
Molecular Formula: C7H8Cl3NO2
InChi Key: GWQBXRYSVSZLSL-UHFFFAOYNA-N
SMILES: ClC(Cl)(Cl)C1OC(=O)C2CCCN12
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)