Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(2R,5S)-2-Trichloromethyl-3-oxa-1-azabicyclo[3.3.0]octan-4-one 98.0+%, TCI America™
SDP

Catalog No. T29021G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T29021G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T29021G Supplier TCI America Supplier No. T29021G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 97538-67-5
Molecular Formula C7H8Cl3NO2
Molecular Weight (g/mol) 244.496
MDL Number MFCD08274464
InChI Key GWQBXRYSVSZLSL-UJURSFKZSA-N
Synonym 3r,7as-3-trichloromethyl tetrahydropyrrolo 1,2-c oxazol-1 3h-one, 2r,5s-2-trichloromethyl-3-oxa-1-azabicyclo 3.3.0 octan-4-one, 3r,7as-3-trichloromethyl-hexahydropyrrolo 1,2-c 1,3 oxazol-1-one, 2r,5s-2-trichloromethyl-3-oxa-1-azabicyclo 3,3,0 octane-4-one, 3r,7as-3-trichloromethyl-tetrahydro-3h-pyrrolo 1,2-c 1,3 oxazol-1-one, l-proline-chloral condensation product, 3r-trichloromethyl-cis-tetrahydropyrrolo 1,2-c oxazol-1-one, 2r,5s-2-trichloromethyl-1-aza-3-oxabicyclo 3.3.0 octan-4-one, 3r,7as-3-trichloromethyl-tetrahydro-pyrrolo 1,2-c oxazol-1-one, 3r,7as-3-trichloromethyl-tetrahydropyrrolo 1,2-c oxazol-1 3h-one
PubChem CID 10944680
IUPAC Name (3R,7aS)-3-(trichloromethyl)-5,6,7,7a-tetrahydro-3H-pyrrolo[1,2-c][1,3]oxazol-1-one
SMILES C1CC2C(=O)OC(N2C1)C(Cl)(Cl)Cl

Specifications

Melting Point 111°C
Color White
Quantity 1 g
Formula Weight 244.50
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material (2R,5S)-2-Trichloromethyl-3-oxa-1-azabicyclo[3.3.0]octan-4-one
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.