CAS RN 97548-97-5
IUPAC Name:
dihydrogen (5aR,9aR)-6-propyl-5H,5aH,6H,7H,8H,9H,9aH,10H-pyrido[2,3-g]quinazolin-2-amine dichloride
Synonyms:
quinelorane
Molecular Weight (g/mol):
319.27
Molecular Formula:
C14H24Cl2N4
InChi Key:
WDEMLQIGYYLRRX-OWVUFADGSA-N
SMILES:
[H+].[H+].[Cl-].[Cl-].CCCN1CCC[C@@H]2CC3=NC(N)=NC=C3C[C@@H]12
1
–
1
of
1
results
1
Tocris Bioscience™ Quinelorane hydrochloride
CAS: 97548-97-5 Molecular Formula: C14H24Cl2N4