accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 976-56-7

IUPAC Name: diethyl [(3,5-di-tert-butyl-4-hydroxyphenyl)methyl]phosphonate
Synonyms: diethyl (3,5-di-tert-butyl-4-hydroxyphenyl)methylphosphonate
Molecular Weight (g/mol): 356.44
Molecular Formula: C19H33O4P
InChi Key: GJDRKHHGPHLVNI-UHFFFAOYSA-N
SMILES: CCOP(=O)(CC1=CC(=C(O)C(=C1)C(C)(C)C)C(C)(C)C)OCC
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results