Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Diethyl 3,5-Di-tert-butyl-4-hydroxybenzylphosphonate 98.0+%, TCI America™
SDP

Catalog No. D29675G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D29675G 5 g
D29671G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D29675G Supplier TCI America Supplier No. D29675G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 976-56-7
Molecular Formula C19H33O4P
Molecular Weight (g/mol) 356.44
MDL Number MFCD00227871
InChI Key GJDRKHHGPHLVNI-UHFFFAOYSA-N
Synonym 3,5-Di-tert-butyl-4-hydroxybenzylphosphonic Acid Diethyl Ester
PubChem CID 70421
IUPAC Name diethyl [(3,5-di-tert-butyl-4-hydroxyphenyl)methyl]phosphonate
SMILES CCOP(=O)(CC1=CC(=C(O)C(=C1)C(C)(C)C)C(C)(C)C)OCC

Specifications

Melting Point 122°C
Color White
Quantity 5 g
Formula Weight 356.44
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Diethyl 3,5-Di-tert-butyl-4-hydroxybenzylphosphonate
EINECSNumber (3)-3376
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.