accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COH3COO

CAS RN 976-71-6

IUPAC Name: 9a,11a-dimethyl-2,3,3a,3b,7,8,9,9a,9b,10,11,11a-dodecahydrospiro[cyclopenta[a]phenanthrene-1,2'-oxolane]-5',7-dione
Synonyms: canrenone
Molecular Weight (g/mol): 340.46
Molecular Formula: C22H28O3
InChi Key: UJVLDDZCTMKXJK-UHFFFAOYNA-N
SMILES: CC12CCC3C(C=CC4=CC(=O)CCC34C)C1CCC21CCC(=O)O1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)