Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Canrenone, 97%, Spectrum™ Chemical
SDP

Catalog No. 18601413 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-601-413 25 g Amber Glass Bottle
18-601-412 5 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-601-413 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. C127425GM
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

C1274, 976-71-6, C22H28O3

Spectrum™ Chemical Canrenone , an aldosterone antagonist, can be used as a diuretic. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.

Chemical Identifiers

CAS 976-71-6
Molecular Formula C22H28O3
Molecular Weight (g/mol) 340.46
InChI Key UJVLDDZCTMKXJK-UHFFFAOYNA-N
IUPAC Name 9a,11a-dimethyl-2,3,3a,3b,7,8,9,9a,9b,10,11,11a-dodecahydrospiro[cyclopenta[a]phenanthrene-1,2'-oxolane]-5',7-dione
SMILES CC12CCC3C(C=CC4=CC(=O)CCC34C)C1CCC21CCC(=O)O1

Specifications

CAS Max % 100%
Ignition Residue 0.1%
Loss on Drying 1%
Packaging Amber Glass Bottle
Quantity 25 g
Percent Purity 97%
Grade Reagent
Chemical Name or Material Canrenone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.