accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOCH2OOOOCH3CH3CH3CH3CH3CH3H3CH3CH3CH3COOOCH3OCH2OOCH3CH3CH3H3C

CAS RN 97600-39-0

IUPAC Name: ethyl 2-{[5,11,17,23-tetra-tert-butyl-26,27,28-tris(2-ethoxy-2-oxoethoxy)pentacyclo[19.3.1.1³,⁷.1⁹,¹³.1¹⁵,¹⁹]octacosa-1(25),3,5,7(28),9(27),10,12,15(26),16,18,21,23-dodecaen-25-yl]oxy}acetate
Synonyms: ethyl 2-{[5,11,17,23-tetra-tert-butyl-26,27,28-tris(2-ethoxy-2-oxoethoxy)pentacyclo[19.3.1.1³,⁷.1⁹,¹³.1¹⁵,¹⁹]octacosa-1(25),3,5,7(28),9(27),10,12,15(26),16,18,21,23-dodecaen-25-yl]oxy}acetate
Molecular Weight (g/mol): 993.29
Molecular Formula: C60H80O12
InChi Key: HZHADWCIBZZJNV-UHFFFAOYSA-N
SMILES: CCOC(=O)COC1=C2CC3=CC(=CC(CC4=C(OCC(=O)OCC)C(CC5=C(OCC(=O)OCC)C(CC1=CC(=C2)C(C)(C)C)=CC(=C5)C(C)(C)C)=CC(=C4)C(C)(C)C)=C3OCC(=O)OCC)C(C)(C)C
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
Physical Form 
  • (2)
Boiling Point 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results