Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Dihexadecyldimethylammonium Bromide

Catalog No. 4202201G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
42-022-01G 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 42-022-01G Supplier Sigma Aldrich Supplier No. 4202201G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Dihexadecyldimethylammonium Bromide

Specifications

CAS 97600-39-0
Melting Point 138°C to 141°C
Boiling Point 34.6°C
Molecular Formula C60H80O12
Linear Formula C60H80O12
MDL Number MFCD00145373
Quantity 250 mg
Synonym Dimethyldihexadecylammonium bromide
Molecular Weight (g/mol) 993.27 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.