accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 97685-00-2

IUPAC Name: 2-(4-{2-[4-(dimethylamino)phenyl]diazen-1-yl}benzenesulfonamido)-3-(1H-indol-3-yl)propanoic acid
Synonyms: 2-(4-{2-[4-(dimethylamino)phenyl]diazen-1-yl}benzenesulfonamido)-3-(1H-indol-3-yl)propanoic acid
Molecular Weight (g/mol): 491.57
Molecular Formula: C25H25N5O4S
InChi Key: SGKGZXUAHWJTIA-UHFFFAOYNA-N
SMILES: CN(C)C1=CC=C(C=C1)N=NC1=CC=C(C=C1)S(=O)(=O)NC(CC1=CNC2=CC=CC=C12)C(O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results