Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dabsyl-L-tryptophan 98.0+%, TCI America™
SDP

Catalog No. D1459100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D1459100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D1459100MG Supplier TCI America Supplier No. D1459100MG

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 97685-00-2
Molecular Formula C25H25N5O4S
Molecular Weight (g/mol) 491.566
MDL Number MFCD00059700
InChI Key SGKGZXUAHWJTIA-DEOSSOPVSA-N
Synonym 4-Dimethylaminoazobenzene-4′C-sulfonyl-L-tryptophan, Dbs-Trp-OH
PubChem CID 14048721
IUPAC Name (2S)-2-[[4-[[4-(dimethylamino)phenyl]diazenyl]phenyl]sulfonylamino]-3-(1H-indol-3-yl)propanoic acid
SMILES CN(C)C1=CC=C(C=C1)N=NC2=CC=C(C=C2)S(=O)(=O)NC(CC3=CNC4=CC=CC=C43)C(=O)O

Specifications

Melting Point 206°C
Quantity 100 mg
Formula Weight 491.57
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Dabsyl-L-tryptophan
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.