accessibility menu, dialog, popup

UserName

Viewing profile as user:
POOOCH3CH3CH3H3C

CAS RN 97739-46-3

IUPAC Name: 1,3,5,7-tetramethyl-8-phenyl-2,4,6-trioxa-8-phosphatricyclo[3.3.1.1³,⁷]decane
Synonyms: 1,3,5,7-tetramethyl-8-phenyl-2,4,6-trioxa-8-phosphatricyclo[3.3.1.1³,⁷]decane
Molecular Weight (g/mol): 292.32
Molecular Formula: C16H21O3P
InChi Key: AVVSJWUWBATQBX-UHFFFAOYSA-N
SMILES: CC12CC3(C)OC(C)(CC(C)(O1)P3C1=CC=CC=C1)O2
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results