Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ 1,3,5,7-Tetramethyl-6-phenyl-2,4,8-trioxa-6-phosphaadamantane

Catalog No. AAH36051ME
Change view
Click to view available options
Quantity:
500 mg
2 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH36051ME 500 mg
AAH3605104 2 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH36051ME Supplier Thermo Scientific™ Supplier No. H36051ME
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 97739-46-3 | C16H21O3P | 292.32 g/mol

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 97739-46-3
Molecular Formula C16H21O3P
Molecular Weight (g/mol) 292.32
MDL Number MFCD10567051
InChI Key AVVSJWUWBATQBX-UHFFFAOYSA-N
Synonym meCgPPh;
IUPAC Name 1,3,5,7-tetramethyl-8-phenyl-2,4,6-trioxa-8-phosphatricyclo[3.3.1.1³,⁷]decane
SMILES CC12CC3(C)OC(C)(CC(C)(O1)P3C1=CC=CC=C1)O2

Specifications

Melting Point 106-111°
Quantity 500 mg
Formula Weight 292.31
Percent Purity 98+%
Physical Form Solid
ShelfLife 5 Years
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.