accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOOONOO

CAS RN 97744-90-6

IUPAC Name: 1-{8-methyl-6-oxo-2H,6H-[1,3]dioxolo[4,5-g]chromen-7-yl}-2,5-dihydro-1H-pyrrole-2,5-dione
Synonyms: 1-{8-methyl-6-oxo-2H-[1,3]dioxolo[4,5-g]chromen-7-yl}pyrrole-2,5-dione
Molecular Weight (g/mol): 299.24
Molecular Formula: C15H9NO6
InChi Key: NSFINZUKLACTRX-UHFFFAOYSA-N
SMILES: CC1=C(N2C(=O)C=CC2=O)C(=O)OC2=CC3=C(OCO3)C=C12
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results